Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Styrene

Catalog No. S4972100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
1 L
4 L
16.5 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S4972100ML 100 mL
S49724L 4 L
S4972165L 16.5 L
S4972500ML 500 mL
S49721L 1 L
S49725ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S4972100ML Supplier Sigma Aldrich Supplier No. S4972100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Styrene

Specifications

CAS 100-42-5
Melting Point -31°C
Density 0.906 g/mL at 25°C
Boiling Point 145°C to 146°C
Molecular Formula C8H8
Refractive Index n20/D 1.546
Linear Formula C6H5CH=CH2
MDL Number MFCD00008612
Quantity 100 mL
Synonym Phenylethylene; Vinylbenzene
Molecular Weight (g/mol) 104.15 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.