Learn More
Medchemexpress LLC Styrene-divinylbenzene sulfonated copolymer | 69011-20-7 | 98% | 25 G
SDP
Catalog No. 5000358505
Encompass_Preferred
Styrene-divinylbenzene sulfonated copolymer is a strongly acidic resin and a cationic exchange resin. It is made from a microporous styrene/divinylbenzene (DVB) co-polymer with a sulfonic acid group.
  • Strongly acidic resin
  • Cationic exchange resin
  • Made from microporous styrene/divinylbenzene (DVB) co-polymer
  • Contains a sulfonic acid group
Catalog No. 50-003-58505 Supplier Medchemexpress LLC Supplier No. 25GHY46846
Styrene-divinylbenzene sulfonated copolymer is a strongly acidic resin and a cationic exchange resin. It is made from a microporous styrene/divinylbenzene (DVB) co-polymer with a sulfonic acid group.
  • Strongly acidic resin
  • Cationic exchange resin
  • Made from microporous styrene/divinylbenzene (DVB) co-polymer
  • Contains a sulfonic acid group