Learn More
Medchemexpress LLC Styrene-divinylbenzene sulfonated copolymer | 69011-20-7 | 98% | 50 G
SDP
Catalog No. 5000358111
Encompass_Preferred
Styrene-divinylbenzene sulfonated copolymer is a strongly acidic resin and a cationic exchange resin. It is manufactured from a microporous styrene/divinylbenzene (DVB) co-polymer that includes a sulfonic acid group.
  • Solid appearance
  • Light yellow to brown color
Catalog No. 50-003-58111 Supplier Medchemexpress LLC Supplier No. HY4684650G
Styrene-divinylbenzene sulfonated copolymer is a strongly acidic resin and a cationic exchange resin. It is manufactured from a microporous styrene/divinylbenzene (DVB) co-polymer that includes a sulfonic acid group.
  • Solid appearance
  • Light yellow to brown color