Learn More
Electron Microscopy Sciences Styrene Monomer: Polyester Resin Kit
SDP
Catalog No. 50980429
Encompass_Preferred
Very low viscosity; monomeric styrene which penetrates rapidly into the tissues. It is soluble in ethanol and acetone. The infiltration and embedding can be done by styrene alone, and there is no need to dilute it with solvents. Polymerization can be done either by UV light (340-400nm) or heat (60 degree C) within 2-3 days without benzoyl peroxide added; 24 hours with 1% benzoyl peroxide added as a catalyst.
Catalog No. 50-980-429 Supplier Electron Microscopy Sciences Supplier No. 14650
Very low viscosity; monomeric styrene which penetrates rapidly into the tissues. It is soluble in ethanol and acetone. The infiltration and embedding can be done by styrene alone, and there is no need to dilute it with solvents. Polymerization can be done either by UV light (340-400nm) or heat (60 degree C) within 2-3 days without benzoyl peroxide added; 24 hours with 1% benzoyl peroxide added as a catalyst.