Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(+)-Styrene Oxide

Catalog No. 5400995G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-009-95G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 54-009-95G Supplier Sigma Aldrich Supplier No. 5400995G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used to produce a homologated epoxide as part of a synthetic approach to (+)-allosedamine.Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.

Specifications

CAS 172035-86-8
Density 0.904 g/mL at 25°C
Boiling Point 65°C
Molecular Formula C4H3IMgS
Linear Formula C4H3IMgS
Refractive Index n20/D 1.534
Quantity 5 g
MDL Number MFCD03427252
Synonym (R)-(+)-Phenyloxirane; (R)-Phenylethylene oxide
Molecular Weight (g/mol) 234.34 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.