Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Styrene, Reagent 450 ML
SDP

Catalog No. 5029149
Encompass_Preferred

(Vinylbenzene)
CAS #100-42-5 C₆H₅CH:CH₂ F.W. 104.14
Assay: >99.0%

A monomeric styrene used as an embedding media for ultramicrotomy. (DeLamater, E.D., et al 1971). A styrene-methacrylate resin embedding method for ultrathin sectioning ( J. Electron Micro., 10:16)

Catalog No. 50-291-49 Supplier Electron Microscopy Sciences Supplier No. 21500
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Vinylbenzene)
CAS #100-42-5 C₆H₅CH:CH₂ F.W. 104.14
Assay: >99.0%

A monomeric styrene used as an embedding media for ultramicrotomy. (DeLamater, E.D., et al 1971). A styrene-methacrylate resin embedding method for ultrathin sectioning ( J. Electron Micro., 10:16)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.