Learn More
Med Vet International StaySafe Self-Sealing Bottle Closures, 28mm, J1080B, 50 Count
Catalog No. 5000495669
Encompass_Preferred
This patented product allows a luer slip syringe to repeatedly withdraw oral liquids from plastic amber pharmacy bottles. The StaySafe is pressed into bottle opening and features flexible sealing rings for compatibilitywith a range of bottle sizes.Size 24 fits snugly into most bottles with necks in its I.D. (inner diameter) range of 20.2mm - 21.4mm.Bottles should be selected for smooth inside necks sizedwith the stated inner diameter range. A smooth neck optimizes the ability of the closure to provide an effective seal.
Catalog No. 50-004-95669 Supplier Med Vet International Supplier No. JOR1080B
This patented product allows a luer slip syringe to repeatedly withdraw oral liquids from plastic amber pharmacy bottles. The StaySafe is pressed into bottle opening and features flexible sealing rings for compatibilitywith a range of bottle sizes.Size 24 fits snugly into most bottles with necks in its I.D. (inner diameter) range of 20.2mm - 21.4mm.Bottles should be selected for smooth inside necks sizedwith the stated inner diameter range. A smooth neck optimizes the ability of the closure to provide an effective seal.