Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SU-4942 | 76086-99-2 | 99.6% | 300.15 | 50 MG
SDP

Catalog No. 5000401670
Encompass_Preferred

SU-4942 is a tyrosine kinase signal modulator that inhibits VEGF- and endothelial cell growth factor (ECGF)-induced mitogenesis in endothelial cells.

  • Tyrosine kinase signal modulator
  • Inhibits VEGF- and ECGF-induced mitogenesis in endothelial cells
  • Solid appearance
  • Light yellow to yellow color
  • Molecular weight: 300.15
  • Purity: 99.6%

Catalog No. 50-004-01670 Supplier Medchemexpress LLC Supplier No. HY2129050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SU-4942 is a tyrosine kinase signal modulator that inhibits VEGF- and endothelial cell growth factor (ECGF)-induced mitogenesis in endothelial cells.

  • Tyrosine kinase signal modulator
  • Inhibits VEGF- and ECGF-induced mitogenesis in endothelial cells
  • Solid appearance
  • Light yellow to yellow color
  • Molecular weight: 300.15
  • Purity: 99.6%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.