Learn More
Medchemexpress LLC SU 5214 | 186611-04-1 | 98.29% | 251.28 | 50 MG
SDP
Catalog No. 5000400674
Encompass_Preferred
SU5214 is a potent VEGFR2 inhibitor extracted from patent US5834504A. It has IC50s of 14.8 μM for FLK-1 and 36.7 μM for EGFR. This product is suitable for various laboratory applications.
  • Potent VEGFR2 inhibitor
  • IC50s of 14.8 μM for FLK-1
  • IC50s of 36.7 μM for EGFR
  • Extracted from patent US5834504A
Catalog No. 50-004-00674 Supplier Medchemexpress LLC Supplier No. HY2129250MG
SU5214 is a potent VEGFR2 inhibitor extracted from patent US5834504A. It has IC50s of 14.8 μM for FLK-1 and 36.7 μM for EGFR. This product is suitable for various laboratory applications.
  • Potent VEGFR2 inhibitor
  • IC50s of 14.8 μM for FLK-1
  • IC50s of 36.7 μM for EGFR
  • Extracted from patent US5834504A