Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Su 5214 | 186611-04-1 | MFCD01595259 | >98.0% | 10 MG
SDP

Catalog No. 5000297019
Encompass_Preferred

SU5214 is a small-molecule tyrosine kinase inhibitor reported to inhibit vascular endothelial growth factor receptor 2 (VEGFR2/FLK-1) and epidermal growth factor receptor (EGFR) in cell-free assays. It is used for biochemical studies and target validation involving VEGFR2/EGFR signaling pathways.

  • Reported IC50 of 14.8 μM for VEGFR2 (cell-free assay).
  • Reported IC50 of 36.7 μM for EGFR (cell-free assay).
  • CAS number 186611-04-1.
  • Molecular weight 251.285 g·mol⁻¹.
  • Purity greater than 98% (HPLC) as supplied by chemical vendors.
  • Supplied in small milligram quantities suitable for in vitro assays.

Catalog No. 50-002-97019 Supplier Medchemexpress LLC Supplier No. HY2129210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SU5214 is a small-molecule tyrosine kinase inhibitor reported to inhibit vascular endothelial growth factor receptor 2 (VEGFR2/FLK-1) and epidermal growth factor receptor (EGFR) in cell-free assays. It is used for biochemical studies and target validation involving VEGFR2/EGFR signaling pathways.

  • Reported IC50 of 14.8 μM for VEGFR2 (cell-free assay).
  • Reported IC50 of 36.7 μM for EGFR (cell-free assay).
  • CAS number 186611-04-1.
  • Molecular weight 251.285 g·mol⁻¹.
  • Purity greater than 98% (HPLC) as supplied by chemical vendors.
  • Supplied in small milligram quantities suitable for in vitro assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.