Learn More
Apexbio Technology LLC SU 5402 215543-92-3 10mg
SDP
Catalog No. 501998376
Encompass_Preferred
SU 5402 (CAS 215543-92-3) is a selective receptor tyrosine kinase inhibitor targeting VEGFR2 FGFR1 and PDGFR with respective IC50 values of 0 02 M 0 03 M and 0 51 M It exhibits minimal inhibitory activity against EGFR ( 100 M IC50) SU 5402 inhibits FGFR-mediated phosphorylation events impacting downstream signaling pathways including ERK1/2 STAT3 and MAP kinases leading to cell cycle arrest and induction of apoptosis particularly in FGFR3-dependent multiple myeloma cell lines This compound serves as a relevant tool for exploring receptor tyrosine kinase signaling in cancer research
Catalog No. 50-199-8376 Supplier Apexbio Technology LLC Supplier No. A384310
SU 5402 (CAS 215543-92-3) is a selective receptor tyrosine kinase inhibitor targeting VEGFR2 FGFR1 and PDGFR with respective IC50 values of 0 02 M 0 03 M and 0 51 M It exhibits minimal inhibitory activity against EGFR ( 100 M IC50) SU 5402 inhibits FGFR-mediated phosphorylation events impacting downstream signaling pathways including ERK1/2 STAT3 and MAP kinases leading to cell cycle arrest and induction of apoptosis particularly in FGFR3-dependent multiple myeloma cell lines This compound serves as a relevant tool for exploring receptor tyrosine kinase signaling in cancer research