Learn More
Medchemexpress LLC SU 5402 10mM 1mL | 215543-92-3 | 10MM 1ML
SDP
Catalog No. 5000243483
Encompass_Preferred
SU 5402 is a potent multi-targeted receptor tyrosine kinase inhibitor with IC50 of 20 nM 30 nM and 510 nM for VEGFR2 FGFR1 and PDGFR respectively
Catalog No. 50-002-43483 Supplier Medchemexpress LLC Supplier No. HY1040710MM 1ML
SU 5402 is a potent multi-targeted receptor tyrosine kinase inhibitor with IC50 of 20 nM 30 nM and 510 nM for VEGFR2 FGFR1 and PDGFR respectively