Learn More
Abcam SU-5402, VEGFR and FGFR inhibitor, 1MG
Catalog No. 502706660
Encompass_Preferred
MW 296.32 Da, Purity >97%. Potent, selective VEGFR and FGFR inhibitor (IC₅₀ values are 0.02 (VEGFR2), 0.03 (FGFR1), 0.51 (PDGFRβ) and >100 μM (EGFR)). Attenuates integrin β4-induced differentiation of neural stem cells. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6660 Supplier Abcam Supplier No. AB1413681MG
MW 296.32 Da, Purity >97%. Potent, selective VEGFR and FGFR inhibitor (IC₅₀ values are 0.02 (VEGFR2), 0.03 (FGFR1), 0.51 (PDGFRβ) and >100 μM (EGFR)). Attenuates integrin β4-induced differentiation of neural stem cells. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement