Learn More
Cayman Chemical SU 5416, 204005-46-9, 50 mg
SDP
Supplier:  Cayman Chemical 1334250
Encompass_Preferred
Molecular Formula C15H14N2O, Purity ≥98%, Formula Weight 238.3, Synonyms: NSC 696819, Semaxinib, Sugen 5416, VEGFR 2 Kinase Inhibitor; A tyrosine kinase inhibitor best known as an inhibitor of VEGFR2 (FLK1/KDR) and a suppressor of tumor vascularization, preventing the growth of multiple tumor types.
Catalog No. NC1561997