Learn More
KAYAKU ADVANCED MATERIALS INC SU-8 3010-500 ML GLASS
Catalog No. NC1248659
Encompass
  • High-contrast photoresist for precise micromachining and microelectronics.
  • Epoxy-based, offering improved adhesion and reduced coating stress.
  • Facilitates high aspect ratio structures with near-vertical sidewalls.
  • Enables film thicknesses from 4 to 100 µm in one coat.
  • Compatible with i-line (365 nm) and low-energy UV (350-400 nm) exposure.
  • High optical transparency above 360 nm enhances imaging quality.
  • Maintains excellent chemical and thermal stability under extreme conditions.
  • Container size of 500 mL in a glass vial.
  • Adhesion is further improved for critical applications.
  • Coating stress is minimized for better performance.
Catalog No. NC1248659 Supplier KAYAKU ADVANCED MATERIALS INC Supplier No. Y311060 0500L1GL
  • High-contrast photoresist for precise micromachining and microelectronics.
  • Epoxy-based, offering improved adhesion and reduced coating stress.
  • Facilitates high aspect ratio structures with near-vertical sidewalls.
  • Enables film thicknesses from 4 to 100 µm in one coat.
  • Compatible with i-line (365 nm) and low-energy UV (350-400 nm) exposure.
  • High optical transparency above 360 nm enhances imaging quality.
  • Maintains excellent chemical and thermal stability under extreme conditions.
  • Container size of 500 mL in a glass vial.
  • Adhesion is further improved for critical applications.
  • Coating stress is minimized for better performance.