Learn More
Enzo Life Sciences SU 9516 (5mg). CAS: 377090-84-1
SDP
Catalog No. 502008536
Encompass_Preferred
Cell permeable, potent, selective and ATP-competitive inhibitor of CDK2/cyclin A (IC50=22nM), CDK1/cyclin B (IC50=40nM) and CDK4/cyclin D1 (IC50=200nM). Has no effect on the activity of PKC, p38, PDGFRβ and EGFR (IC50>10µM). Displays anti-proliferative and pro-apoptotic properties in tumor cells. Inhibits proliferation of growth factor-stimulated colon carcinoma cells by binding to CDK2 and thus preventing the phosphorylation of pRb and its dissociation from E2F. Alternative name: 3-[1-(3H-Imidazol-4-yl)-meth-(Z)-ylidene]-5-methoxy-1,3-dihydro-indol-2-one. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-8536 Supplier Enzo Life Sciences Supplier No. ALX270400M005
Cell permeable, potent, selective and ATP-competitive inhibitor of CDK2/cyclin A (IC50=22nM), CDK1/cyclin B (IC50=40nM) and CDK4/cyclin D1 (IC50=200nM). Has no effect on the activity of PKC, p38, PDGFRβ and EGFR (IC50>10µM). Displays anti-proliferative and pro-apoptotic properties in tumor cells. Inhibits proliferation of growth factor-stimulated colon carcinoma cells by binding to CDK2 and thus preventing the phosphorylation of pRb and its dissociation from E2F. Alternative name: 3-[1-(3H-Imidazol-4-yl)-meth-(Z)-ylidene]-5-methoxy-1,3-dihydro-indol-2-one. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO (10mg/ml). Long Term Storage: +4°C.