Learn More
Foxx Life Sciences EZBio MTO Single Use (SUT) Bottle Assembly, 2000mL (2L) Sterilized PETG Bottle for Aseptic Sampling and Sterile Storage, 53B, EZBio Tubing, Vented w/ Diptube, CPC MPC, 10/CS
GREENER_CHOICE
Catalog No. 502060784
Encompass_Preferred
Made To Order MTO Single Use Sterile 2000mL (2L) PETG Media Bottle Assembly w/ Tubing and Vent Filters. EZBio MTO Single Use Media Bottle Assemblies include a 2-port 53B VersaCap assembly with 1/8" ID (3.2mm) x 1/4" OD (6.4mm) EZBio® TPE Tubing and diptube. This bottle assembly features a CPC MPC connector that allows for quick connections and the ability to transfer media easily with less risk of error. Additionally, all bottle assemblies include a 0.2µm Hydrophobic PTFE Vent Filter that allows for ventilation while preventing media from escaping in the event of a spill. EZBio "MTO" Single Use Media Bottle Assemblies also include an extra closed cap for storage and a ratchet style pinch clamp that allows the user to control flow into the container or close off the line entirely. The EZBio "MTO" Bottle Assembly is the perfect option to phase out costly stainless steel apparatuses that are expensive to clean and validate. USP Class VI components, Gamma Irradiated in USA.
Catalog No. 50-206-0784 Supplier Foxx Life Sciences Supplier No. 4370107FLS
Made To Order MTO Single Use Sterile 2000mL (2L) PETG Media Bottle Assembly w/ Tubing and Vent Filters. EZBio MTO Single Use Media Bottle Assemblies include a 2-port 53B VersaCap assembly with 1/8" ID (3.2mm) x 1/4" OD (6.4mm) EZBio® TPE Tubing and diptube. This bottle assembly features a CPC MPC connector that allows for quick connections and the ability to transfer media easily with less risk of error. Additionally, all bottle assemblies include a 0.2µm Hydrophobic PTFE Vent Filter that allows for ventilation while preventing media from escaping in the event of a spill. EZBio "MTO" Single Use Media Bottle Assemblies also include an extra closed cap for storage and a ratchet style pinch clamp that allows the user to control flow into the container or close off the line entirely. The EZBio "MTO" Bottle Assembly is the perfect option to phase out costly stainless steel apparatuses that are expensive to clean and validate. USP Class VI components, Gamma Irradiated in USA.