Learn More
Foxx Life Sciences EZBio MTO, Single Use (SUT) Bottle Assembly, 2000mL (2L) Sterilized PETG Bottle for Aseptic Sampling and Sterile Storage, 53B, Flexelene®, Vented w/ Diptube, Plug, 10/CS
GREENER_CHOICE
Catalog No. 502060491
Encompass_Preferred
Made To Order MTO Single Use Sterile 2000mL (2L) PETG Media Bottle Assemblies with Tubing and Vent Filters. EZBio MTO Single Use Media Bottle Assemblies include a 2-port 53B VersaCap assembly with 1/8" ID (3.2mm) x 1/4" OD (6.4mm) Flexelene® TPE Tubing and diptube. This bottle assembly features a hose barb plug that keeps the line closed until it is removed for use. Additionally, all bottle assemblies include a 0.2µm Hydrophobic PTFE Vent Filter that allows for ventilation while preventing media from escaping in the event of a spill. All EZBio "MTO" Single Use Media Bottle Assemblies also include an extra closed cap for storage and a ratchet style pinch clamp that allows the user to control flow into the container or close off the line entirely. The EZBio "MTO" Bottle Assembly is the perfect option to phase out costly stainless steel apparatuses that are expensive to clean and validate. USP Class VI components, Gamma Irradiated in USA.
Catalog No. 50-206-0491 Supplier Foxx Life Sciences Supplier No. 4374007FLS
Made To Order MTO Single Use Sterile 2000mL (2L) PETG Media Bottle Assemblies with Tubing and Vent Filters. EZBio MTO Single Use Media Bottle Assemblies include a 2-port 53B VersaCap assembly with 1/8" ID (3.2mm) x 1/4" OD (6.4mm) Flexelene® TPE Tubing and diptube. This bottle assembly features a hose barb plug that keeps the line closed until it is removed for use. Additionally, all bottle assemblies include a 0.2µm Hydrophobic PTFE Vent Filter that allows for ventilation while preventing media from escaping in the event of a spill. All EZBio "MTO" Single Use Media Bottle Assemblies also include an extra closed cap for storage and a ratchet style pinch clamp that allows the user to control flow into the container or close off the line entirely. The EZBio "MTO" Bottle Assembly is the perfect option to phase out costly stainless steel apparatuses that are expensive to clean and validate. USP Class VI components, Gamma Irradiated in USA.