Learn More
Foxx Life Sciences EZBio MTO Single Use (SUT) Cap Assembly, 38-430 VersaCap, PP, C-Flex Tubing, Vented w/ Diptube, Plug, for 1000 mL (1L) Bottle, Foxx or Nalgene Bottles, Sterile, 20/CS
Catalog No. 502060880
Encompass_Preferred
Made To Order MTO Sterile Single-Use Polypropylene Cap Assembly for 1000mL (1L) Bottles with Tubing and Vent Filters are perfect for fluid transfer and storage. This EZBio MTO Single Use VersaCap assembly features a 2-port 38-430 Cap with 1/8” ID (3.2mm) x ¼” OD (6.4mm) C-Flex® TPE Tubing and diptube. This cap assembly features a hose barb plug that keeps the line closed until it is removed for use. Additionally, all cap assemblies include a 0.2µm Hydrophobic PTFE Vent Filter that allows for ventilation while preventing media from escaping in the event of a spill. All EZBio "MTO" Single Use Cap Assemblies also include a ratchet style pinch clamp that allows the user to control flow into their container or close off the line entirely. The EZBio "MTO" Cap Assembly is the perfect option to phase out costly stainless steel apparatuses that are expensive to clean and validate. USP Class VI components, Gamma Irradiated in USA.
Catalog No. 50-206-0880 Supplier Foxx Life Sciences Supplier No. 4301017FLS
Made To Order MTO Sterile Single-Use Polypropylene Cap Assembly for 1000mL (1L) Bottles with Tubing and Vent Filters are perfect for fluid transfer and storage. This EZBio MTO Single Use VersaCap assembly features a 2-port 38-430 Cap with 1/8” ID (3.2mm) x ¼” OD (6.4mm) C-Flex® TPE Tubing and diptube. This cap assembly features a hose barb plug that keeps the line closed until it is removed for use. Additionally, all cap assemblies include a 0.2µm Hydrophobic PTFE Vent Filter that allows for ventilation while preventing media from escaping in the event of a spill. All EZBio "MTO" Single Use Cap Assemblies also include a ratchet style pinch clamp that allows the user to control flow into their container or close off the line entirely. The EZBio "MTO" Cap Assembly is the perfect option to phase out costly stainless steel apparatuses that are expensive to clean and validate. USP Class VI components, Gamma Irradiated in USA.