Learn More
Pendotech Pendotech 1/4 Inch Hose Barb Single Use Conductivity Sensor
Catalog No. NC0911828
Encompass
Single Use Conductivity Sensor, non-sterile, polysulfone 1/4 inch hose barb
This item is not returnable. View return policy
Catalog No. NC0911828 Supplier Pendotech Supplier No. CONDSN025
Single Use Conductivity Sensor, non-sterile, polysulfone 1/4 inch hose barb