Learn More
CARR BIOSYSTEMS SU MOD UFMINI1/4TPE1/8 IRR
Catalog No. NC3371544
Encompass
  • Single-use module for various bioprocessing applications
  • Compatible with UFMini single-use centrifuge system
  • Gamma irradiated for sterility
  • Endotoxin-free environment
  • Suitable for cell clarification, harvesting, washing, and concentration
  • Utilizes low-shear tubular bowl centrifugation
  • Ensures high cell viability
  • Maximizes product recovery
  • Free from animal-derived components
  • Module inlet/outlet designation: 1/8 in
Catalog No. NC3371544 Supplier CARR BIOSYSTEMS Supplier No. C68390007
  • Single-use module for various bioprocessing applications
  • Compatible with UFMini single-use centrifuge system
  • Gamma irradiated for sterility
  • Endotoxin-free environment
  • Suitable for cell clarification, harvesting, washing, and concentration
  • Utilizes low-shear tubular bowl centrifugation
  • Ensures high cell viability
  • Maximizes product recovery
  • Free from animal-derived components
  • Module inlet/outlet designation: 1/8 in