Learn More
World Precision Instrument SU-MPC385 Motorized Micromanipulator Systems-505089
SDP
Catalog No. 502539254
Encompass
  • Quietest electronics in the industry optimized for single channel recording
  • Single controller and ROE will run two stepper motor drive manipulators
  • Self-detecting, daisy-chain capability allows control of four manipulators from one 505093
  • User-friendly interface: single button access to all major functions
  • Easily configurable virtual 4th axis set without external computer
  • Accelerated Mode for fast, manual manipulator movement
  • Easy toggle selection of Mode (speed/resolution, pulsed diagonal, Accelerated Mode)
  • Display indicates X, Y, Z coordinates, Mode, active manipulator
  • Robotic Home and Work Position moves for easy automated pipette exchange
  • Faster robotic moves than previous versions
  • Definable 4th axis for coaxial pipette movement, angle selected by DIP switches
  • Simple USB interface
Catalog No. 50-253-9254 Supplier World Precision Instrument Supplier No. 505089
Add to Cart
Add to Cart
Request a Quote
  • Quietest electronics in the industry optimized for single channel recording
  • Single controller and ROE will run two stepper motor drive manipulators
  • Self-detecting, daisy-chain capability allows control of four manipulators from one 505093
  • User-friendly interface: single button access to all major functions
  • Easily configurable virtual 4th axis set without external computer
  • Accelerated Mode for fast, manual manipulator movement
  • Easy toggle selection of Mode (speed/resolution, pulsed diagonal, Accelerated Mode)
  • Display indicates X, Y, Z coordinates, Mode, active manipulator
  • Robotic Home and Work Position moves for easy automated pipette exchange
  • Faster robotic moves than previous versions
  • Definable 4th axis for coaxial pipette movement, angle selected by DIP switches
  • Simple USB interface