Learn More
World Precision Instrument SU-P2000 Micropipette Puller
SDP
Catalog No. 502538516
Encompass
The SU-P2000 micropipette puller represents a significant advance in the technology of fabrication of micropipettes, optical fiber probes, and nanospray tips. The P-2000 integrates a CO2 laser-based heat source with the technology derived from our extensive experience with conventional pullers. This system offers capabilities unmatched by other pullers.
View our Micropipette Puller Comparison Chart
Catalog No. 50-253-8516 Supplier World Precision Instrument Supplier No. SUP2000
Add to Cart
Add to Cart
Request a Quote
The SU-P2000 micropipette puller represents a significant advance in the technology of fabrication of micropipettes, optical fiber probes, and nanospray tips. The P-2000 integrates a CO2 laser-based heat source with the technology derived from our extensive experience with conventional pullers. This system offers capabilities unmatched by other pullers.
View our Micropipette Puller Comparison Chart