Learn More
Pendotech Pendotech 3/4 Inch Hose Barb Single Use Pressure Sensor
Catalog No. NC1360931
Encompass
This item has a minimum order quantity of 5 per supplier requirements
Single use UV flow cell, 2mm path length, non-sterile, polysulfone, 1/8" inch hose barb
Catalog No. NC1360931 Supplier Pendotech Supplier No. PREPSN075
This item has a minimum order quantity of 5 per supplier requirements
Single use UV flow cell, 2mm path length, non-sterile, polysulfone, 1/8" inch hose barb