Learn More
TARGETMOL CHEMICALS INC SU11274 10MG
SDP
Catalog No. 502579512
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SU11274 (Met Kinase Inhibitor)(IC50=10 nM) is a specific Met inhibitor. It shows no significant effects on PGDFR(beta) EGFR or Tie2. purity: 98%
Catalog No. 50-257-9512 Supplier TARGETMOL CHEMICALS INC Supplier No. T615410MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SU11274 (Met Kinase Inhibitor)(IC50=10 nM) is a specific Met inhibitor. It shows no significant effects on PGDFR(beta) EGFR or Tie2. purity: 98%