Learn More
Apexbio Technology LLC SU11274, 5mg. Cas: 658084-23-2 MFCD: MFCD08276928. C-Met inhibitor,potent and selective
SDP
Catalog No. 501904759
Encompass_Preferred
SU11274 is a potent and selective inhibitor of Met kinase with IC50 value of 10nM [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4759 Supplier Apexbio Technology LLC Supplier No. A26785
SU11274 is a potent and selective inhibitor of Met kinase with IC50 value of 10nM [1]. Other sizes are available. Please inqury us for quote.