Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Su11274 (PKI-SU11274) | 658084-23-2 | 98.8% | 5 MG
SDP

Catalog No. 5000232076
Encompass_Preferred

SU11274 is a small-molecule selective inhibitor of the c-Met (HGFR) receptor tyrosine kinase with a reported IC50 of 10 nM. It is supplied as a high-purity research reagent suitable for biochemical and cellular studies of Met-driven signaling pathways.

  • Selective c-Met inhibitor with reported IC50 of 10 nM.
  • High purity, approximately 98.8% (HPLC).
  • Molecular formula C28H30ClN5O4S.
  • Molecular weight 568.1 g/mol.
  • Available in multiple pack sizes, including 5 mg.
  • Safety data sheet available for handling and storage information.

Catalog No. 50-002-32076 Supplier Medchemexpress LLC Supplier No. HY120145MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SU11274 is a small-molecule selective inhibitor of the c-Met (HGFR) receptor tyrosine kinase with a reported IC50 of 10 nM. It is supplied as a high-purity research reagent suitable for biochemical and cellular studies of Met-driven signaling pathways.

  • Selective c-Met inhibitor with reported IC50 of 10 nM.
  • High purity, approximately 98.8% (HPLC).
  • Molecular formula C28H30ClN5O4S.
  • Molecular weight 568.1 g/mol.
  • Available in multiple pack sizes, including 5 mg.
  • Safety data sheet available for handling and storage information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.