Learn More
Selleck Chemical LLC SU11274 25mg 658084-23-2 PKI-SU11274
SDP
Catalog No. 508526
Encompass_Preferred
SU11274 (PKI-SU11274) is a selective Met (c-Met) inhibitor with IC50 of 10 nM in cell-free assays, no effects on PGDFR?, EGFR or Tie2. SU11274 induces autophagy, apoptosis and cell cycle arrest. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-852-6 Supplier Selleck Chemical LLC Supplier No. S108025MG
SU11274 (PKI-SU11274) is a selective Met (c-Met) inhibitor with IC50 of 10 nM in cell-free assays, no effects on PGDFR?, EGFR or Tie2. SU11274 induces autophagy, apoptosis and cell cycle arrest. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.