Learn More
Medchemexpress LLC HY-10501A 5mg Medchemexpress, SU14813 (maleate) CAS:849643-15-8 Purity:>98%
SDP
Catalog No. 501968241
Encompass_Preferred
Medchemexpress, HY-10501A 5mg SU14813 (maleate) CAS:849643-15-8 SU14813 maleate is a multi-targeted receptor tyrosine kinases inhibitor with IC50s of 50, 2, 4, 15 nM for VEGFR2, VEGFR1, PDGFRβ and KIT. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8241 Supplier Medchemexpress LLC Supplier No. HY10501A5MG
Medchemexpress, HY-10501A 5mg SU14813 (maleate) CAS:849643-15-8 SU14813 maleate is a multi-targeted receptor tyrosine kinases inhibitor with IC50s of 50, 2, 4, 15 nM for VEGFR2, VEGFR1, PDGFRβ and KIT. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.