Learn More
Medchemexpress LLC SU14813 maleate | 849643-15-8 | MFCD24682713 | 99.0% | 558.56 | C27H31FN4O8 | 10 MG
SDP
Catalog No. 5000243619
Encompass_Preferred
SU14813 maleate is a multi-targeted receptor tyrosine kinase inhibitor provided for research use. It shows low-nanomolar inhibitory activity against VEGFR2, VEGFR1, PDGFRβ, and KIT, and is supplied as a solid or as a 10 mM solution in DMSO for laboratory studies.
  • Multi-target RTK inhibition with low-nanomolar potency.
  • Activity data reported for VEGFR2, VEGFR1, PDGFRβ, and KIT.
  • Available as solid packs and a 10 mM DMSO solution for easy preparation.
  • High reported purity appropriate for in vitro and preclinical research.
  • Supplied with SDS documentation for safe laboratory handling.
Catalog No. 50-002-43619 Supplier Medchemexpress LLC Supplier No. HY10501A10MG
SU14813 maleate is a multi-targeted receptor tyrosine kinase inhibitor provided for research use. It shows low-nanomolar inhibitory activity against VEGFR2, VEGFR1, PDGFRβ, and KIT, and is supplied as a solid or as a 10 mM solution in DMSO for laboratory studies.
  • Multi-target RTK inhibition with low-nanomolar potency.
  • Activity data reported for VEGFR2, VEGFR1, PDGFRβ, and KIT.
  • Available as solid packs and a 10 mM DMSO solution for easy preparation.
  • High reported purity appropriate for in vitro and preclinical research.
  • Supplied with SDS documentation for safe laboratory handling.