Learn More
Medchemexpress LLC SU14813 maleate | 849643-15-8 | MFCD24682713 | 99.0% | 558.56 g/mol | C27H31FN4O8 | 50 MG
SDP
Catalog No. 5000244207
Encompass_Preferred
SU14813 maleate is the maleate salt of SU14813, a small-molecule multi-targeted receptor tyrosine kinase inhibitor with nanomolar potency against VEGFR1, VEGFR2, PDGFRβ, and KIT. It is supplied for research use and characterized by a defined chemical formula and high purity suitable for biochemical and cellular studies.
  • Potent inhibition of VEGFR1 (IC50 2 nM), VEGFR2 (IC50 50 nM), PDGFRβ (IC50 4 nM), and KIT (IC50 15 nM).
  • Maleate salt form to aid solubility and handling in aqueous systems.
  • High purity suitable for research-grade experiments.
  • Applicable to biochemical kinase assays and cellular target validation studies.
  • Provided in small, laboratory-scale quantities for research applications.
Catalog No. 50-002-44207 Supplier Medchemexpress LLC Supplier No. HY10501A50MG
SU14813 maleate is the maleate salt of SU14813, a small-molecule multi-targeted receptor tyrosine kinase inhibitor with nanomolar potency against VEGFR1, VEGFR2, PDGFRβ, and KIT. It is supplied for research use and characterized by a defined chemical formula and high purity suitable for biochemical and cellular studies.
  • Potent inhibition of VEGFR1 (IC50 2 nM), VEGFR2 (IC50 50 nM), PDGFRβ (IC50 4 nM), and KIT (IC50 15 nM).
  • Maleate salt form to aid solubility and handling in aqueous systems.
  • High purity suitable for research-grade experiments.
  • Applicable to biochemical kinase assays and cellular target validation studies.
  • Provided in small, laboratory-scale quantities for research applications.