Learn More
Enzo Life Sciences SU4312 (25mg). CAS: 5812-07-7
SDP
Supplier:  Enzo Life Sciences BMLEI3060025
Encompass_Preferred
Potent and selective inhibitor of VEGFR-2 (Flk-1, KDR). It displays a 100-fold greater potency for the unactivated form of the kinase (Ki=0.04µM) compared to the activated form (Ki=4µM). Purity: ≥98%. Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0691