Learn More
Medchemexpress LLC HY-126319 5mg Medchemexpress, SU5204 CAS:186611-11-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1263195MG
Encompass_Preferred
Medchemexpress, HY-126319 5mg SU5204 CAS:186611-11-0 SU5204, a tyrosine kinase inhibitor, has IC50s of 4 and 51.5 μM for FLK-1 (VEGFR-2) and HER2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1096