Learn More
Medchemexpress LLC SU5205 | 3476-86-6 | 98.02% | 239.24 | 50 MG
SDP
Catalog No. 5000401392
Encompass_Preferred
SU5205 is an inhibitor of VEGFR2 (FLK-1), with an IC50 of 9.6 μM. It inhibits ligand-induced endothelial mitogenesis for VEGF with an IC50 of 5.1 μM.
  • Inhibitor of VEGFR2 (FLK-1)
  • Inhibits ligand-induced endothelial mitogenesis for VEGF
Catalog No. 50-004-01392 Supplier Medchemexpress LLC Supplier No. HY2128950MG
SU5205 is an inhibitor of VEGFR2 (FLK-1), with an IC50 of 9.6 μM. It inhibits ligand-induced endothelial mitogenesis for VEGF with an IC50 of 5.1 μM.
  • Inhibitor of VEGFR2 (FLK-1)
  • Inhibits ligand-induced endothelial mitogenesis for VEGF