Learn More
Medchemexpress LLC SU5214 | 186611-04-1 | 251.28 | 100 MG
SDP
Catalog No. 5000402123
Encompass_Preferred
SU5214 is a potent VEGFR2 inhibitor. It is extracted from patent US5834504A and is suitable for research use. It acts on Protein Tyrosine Kinase/RTK pathway.
  • Potent VEGFR2 inhibitor
  • Target for VEGFR
  • Inhibits FLK-1 with an IC50 of 14.8 μM
  • Inhibits EGFR with an IC50 of 36.7 μM
Catalog No. 50-004-02123 Supplier Medchemexpress LLC Supplier No. HY21292100MG
SU5214 is a potent VEGFR2 inhibitor. It is extracted from patent US5834504A and is suitable for research use. It acts on Protein Tyrosine Kinase/RTK pathway.
  • Potent VEGFR2 inhibitor
  • Target for VEGFR
  • Inhibits FLK-1 with an IC50 of 14.8 μM
  • Inhibits EGFR with an IC50 of 36.7 μM