Learn More
STEMCELL Technologies SU5402, Size: 5 mg
SDP
Supplier:  STEMCELL Technologies 73914
Encompass
SU5402 is a potent and selective inhibitor of vascular endothelial growth factor receptor 2 (VEGFR-2), fibroblast growth factor receptor 1 (FGFR-1), and platelet-derived growth factor receptor beta (PDGFRB) with IC50 = 0.02, 0.03, and 0.51 µM, respectively (Sun et al.).
Catalog No. 50-197-6148
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov