Learn More
Medchemexpress LLC HY-103002 1mg Medchemexpress, SU5408 CAS:15966-93-5 Purity:>98%
SDP
Catalog No. 501968008
Encompass_Preferred
Medchemexpress, HY-103002 1mg SU5408 CAS:15966-93-5 SU5408 (VEGFR2 Kinase Inhibitor I) is a potent and cell-permeable inhibitor of VEGFR2 kinase with an IC50 of 70 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8008 Supplier Medchemexpress LLC Supplier No. HY1030021MG
Medchemexpress, HY-103002 1mg SU5408 CAS:15966-93-5 SU5408 (VEGFR2 Kinase Inhibitor I) is a potent and cell-permeable inhibitor of VEGFR2 kinase with an IC50 of 70 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.