Learn More
TARGETMOL CHEMICALS INC SU5408 5MG
SDP
Catalog No. 502579998
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SU5408 (VEGFR2 Kinase Inhibitor I) is a potent cell-permeable inhibitor of the VEGFR2 kinase. purity: 98%
Catalog No. 50-257-9998 Supplier TARGETMOL CHEMICALS INC Supplier No. T40265MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SU5408 (VEGFR2 Kinase Inhibitor I) is a potent cell-permeable inhibitor of the VEGFR2 kinase. purity: 98%