Learn More
Medchemexpress LLC HY-10374 100mg , SU5416 Semaxinib CAS:204005-46-9 Purity:98%
SDP
Catalog No. 501492674
Encompass_Preferred
Medchemexpress, HY-10374 100mg SU5416 Semaxinib CAS:204005-46-9 Formula:C15H14N2O IC50: 1.23±0.2 μM (Flk-1 receptor) Purity:98% SU5416 is a potent and selective inhibitor of the VEGFR ( Flk-1 / KDR ) that inhibits tyrosine kinase catalysis, and inhibits autophosphorylation of the Flk-1 receptor with an IC 50 of 1.23±0.2 μM (n=4) using an ELISA-based biochemical kinase assay. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2674 Supplier Medchemexpress LLC Supplier No. HY10374100MG
Medchemexpress, HY-10374 100mg SU5416 Semaxinib CAS:204005-46-9 Formula:C15H14N2O IC50: 1.23±0.2 μM (Flk-1 receptor) Purity:98% SU5416 is a potent and selective inhibitor of the VEGFR ( Flk-1 / KDR ) that inhibits tyrosine kinase catalysis, and inhibits autophosphorylation of the Flk-1 receptor with an IC 50 of 1.23±0.2 μM (n=4) using an ELISA-based biochemical kinase assay. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.