Learn More
BIOGEMS INTERNATIONAL INC SU5416 50mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 204469050MG
Encompass
SU5416 is a potent and selective inhibitor of the Flk-1/KDR vascular endothelial growth factor (VEGF) receptor tyrosine kinase and also inhibits c-kit, RET, and FLT3. It is reported to inhibit tumor vascularization and suppress the growth of multiple tumor types.
Catalog No. 50-152-9921