Learn More
Apexbio Technology LLC SU5416, 204005-46-9, MFCD09763655, 50 mg
SDP
Supplier:  Apexbio Technology LLC A384750
Encompass_Preferred
MF: C15H14N2O, MW: 238.28, Synonyms: Semaxinib; SU-5416; SU 5416. Solubility: 11.9 mg/mL in DMSO, <2.28 mg/mL in EtOH, <2.38 mg/mL in H2O. SU5416 was found to inhibit VEGF-dependent phosphorylation of the Flk-1 receptor in Flk-1-overexpressing NIH 3T3 cells.
Catalog No. 50-101-3315