Learn More
Apexbio Technology LLC SU6656 330161-87-0 10mM (in 1mL DMSO)
SDP
Catalog No. 501152208
Encompass_Preferred
SU6656 (CAS 330161-87-0) is a selective small-molecule inhibitor targeting Src family tyrosine kinases enzymes involved in pathways promoting cellular survival proliferation angiogenesis and invasion In NIH 3T3 cells SU6656 suppresses PDGF- and Src-induced cell cycle progression and c-Myc expression In UT-7/TPO leukemia cells SU6656 induces cell cycle arrest with increased DNA content and elevates CD41/CD61 surface expression Additionally SU6656 enhances radiation-mediated endothelial apoptosis and vascular disruption by inhibiting Src-dependent Akt phosphorylation suggesting its potential utility in antitumor angiogenesis research
Catalog No. 50-115-2208 Supplier Apexbio Technology LLC Supplier No. B58395.1
 Restricted Item
Add to Cart
Add to Cart
SU6656 (CAS 330161-87-0) is a selective small-molecule inhibitor targeting Src family tyrosine kinases enzymes involved in pathways promoting cellular survival proliferation angiogenesis and invasion In NIH 3T3 cells SU6656 suppresses PDGF- and Src-induced cell cycle progression and c-Myc expression In UT-7/TPO leukemia cells SU6656 induces cell cycle arrest with increased DNA content and elevates CD41/CD61 surface expression Additionally SU6656 enhances radiation-mediated endothelial apoptosis and vascular disruption by inhibiting Src-dependent Akt phosphorylation suggesting its potential utility in antitumor angiogenesis research