Learn More
REPLIGEN CORPORATION Repligen SUATF6-PES-AQL, Single-Use Design, Low Shear Profile, 50 – 200 L
SDP
Catalog No. NC3345070
Encompass
The Repligen SUATF6-PES-AQL is a single-use cell retention device optimized for high-density cell culture applications. Its diaphragm pumping mechanism ensures low shear stress, making it ideal for efficient cell retention in perfusion and fed-batch processes. This pre-sterilized device offers quick implementation and easy scalability, streamlining bioprocessing while eliminating autoclaving needs. Compatible with various controllers, it enables seamless integration into existing workflows.
Catalog No. NC3345070 Supplier REPLIGEN CORPORATION Supplier No. SUATF6PESAQL
Add to Cart
Add to Cart
Request a Quote
The Repligen SUATF6-PES-AQL is a single-use cell retention device optimized for high-density cell culture applications. Its diaphragm pumping mechanism ensures low shear stress, making it ideal for efficient cell retention in perfusion and fed-batch processes. This pre-sterilized device offers quick implementation and easy scalability, streamlining bioprocessing while eliminating autoclaving needs. Compatible with various controllers, it enables seamless integration into existing workflows.