Learn More
Bio Rad Laboratories SUB-CELL GT CELL WIDE MINI R
Catalog No. 509011328
Encompass
GT Cell; Bio-Rad; Wide Mini ReadySub-Cell; Includes buffer tank, lid and electrodes, leveling bubble; accommodates 20-well, 32-well, and 2 x 32-well wide mini ReadyAgarose gels
Catalog No. 50-901-1328 Supplier Bio Rad Laboratories Supplier No. 1704489
GT Cell; Bio-Rad; Wide Mini ReadySub-Cell; Includes buffer tank, lid and electrodes, leveling bubble; accommodates 20-well, 32-well, and 2 x 32-well wide mini ReadyAgarose gels