Learn More
Bio Rad Laboratories SUB-CELL GT SYSTEM
Catalog No. 509011726
Encompass
System; Bio-Rad; Sub-Cell; UV-transparent gel trays; Gel-casting gates; Replaceable electrode cassettes; Multichannel-pipet; Fixed height drop-in combs; Buffer recirculation ports; With 15 x 20cm tray, gel caste
Catalog No. 50-901-1726 Supplier Bio Rad Laboratories Supplier No. 1704483
System; Bio-Rad; Sub-Cell; UV-transparent gel trays; Gel-casting gates; Replaceable electrode cassettes; Multichannel-pipet; Fixed height drop-in combs; Buffer recirculation ports; With 15 x 20cm tray, gel caste