Learn More
Bio Rad Laboratories Sub-Cell GT UV-Transparent Gel Tray
Catalog No. NC1369164
Encompass
The Sub-Cell GT UV-Transparent Gel Tray is 15x10cm and has two slots for fixed-height combs. It is for use with Wide Mini-Sub Cell GT and Sub-Cell GT Systems.
Catalog No. NC1369164 Supplier Bio Rad Laboratories Supplier No. 1704416
The Sub-Cell GT UV-Transparent Gel Tray is 15x10cm and has two slots for fixed-height combs. It is for use with Wide Mini-Sub Cell GT and Sub-Cell GT Systems.