Learn More
Hoefer, Inc SUB Evo Horizontal Electrophoresis System
SDP
Catalog No. NC3942888
Encompass_Preferred
Designed for high-throughput nucleic acid analysis, can process up to two 96-well plates at once and works seamlessly with 8 or 12-channel pipettes for fast, efficient sample loading. The plug-free lid makes operation easier while protecting the electrodes. With a multi-functional casting stand, three gel tray sizes, and six compatible combs, you can speed up the workflow from gel casting to sample loading.Features and BenefitsRun up to 216 samplesSequential 8-Channel Loading Flexible Gel Setup UV-Transparent Tray Safe & Portable DesignSpecificationsGel Dimensions (w x l): 17 x 25 cm, 17 x 17.5 cm, and 17x 7.5 cmComb: 1.5 mm, 18+36 wellsBase Buffer: 900 mLMaximum Power Settings: 1000 V, 500 mA, 500 WSample Capacity: 216Unit Dimensions (l x w x h): 42.2 x 19.7 x 7.2 cmSafety Certifications: CEIncludes:Large gel tray, 17 x 27 cm, 1 pcs Medium gel tray, 17 x 17.5 cm, 1 pcsMini gel trays, 17 x 7.5 cm, 1 pcsLarge comb 18+36 wells, 1.5 mm thick, 6 pcsCasting stand
Catalog No. NC3942888 Supplier Hoefer, Inc Supplier No. SUB EVO
Add to Cart
Add to Cart
Designed for high-throughput nucleic acid analysis, can process up to two 96-well plates at once and works seamlessly with 8 or 12-channel pipettes for fast, efficient sample loading. The plug-free lid makes operation easier while protecting the electrodes. With a multi-functional casting stand, three gel tray sizes, and six compatible combs, you can speed up the workflow from gel casting to sample loading.Features and BenefitsRun up to 216 samplesSequential 8-Channel Loading Flexible Gel Setup UV-Transparent Tray Safe & Portable DesignSpecificationsGel Dimensions (w x l): 17 x 25 cm, 17 x 17.5 cm, and 17x 7.5 cmComb: 1.5 mm, 18+36 wellsBase Buffer: 900 mLMaximum Power Settings: 1000 V, 500 mA, 500 WSample Capacity: 216Unit Dimensions (l x w x h): 42.2 x 19.7 x 7.2 cmSafety Certifications: CEIncludes:Large gel tray, 17 x 27 cm, 1 pcs Medium gel tray, 17 x 17.5 cm, 1 pcsMini gel trays, 17 x 7.5 cm, 1 pcsLarge comb 18+36 wells, 1.5 mm thick, 6 pcsCasting stand