Learn More
Starna Cells Inc SUB MICRO CELL 100UL
SDP
Catalog No. NC9985694
Encompass_Preferred
Sub-Micro Spectrophotometer Cells, Clear windows: 2, Window Material: Spectrosil Quartz, Useable Range: 170 to 2700nm, Pathlength: 10mm, Aqueous Volume: 115 microliter, Exterior: 12.5L x 12.5W x 45mmH, Window: 2W x 5mmH, Z-Dimension: 15mm
Catalog No. NC9985694 Supplier Starna Cells Inc Supplier No. 16.100Q10/Z15
Sub-Micro Spectrophotometer Cells, Clear windows: 2, Window Material: Spectrosil Quartz, Useable Range: 170 to 2700nm, Pathlength: 10mm, Aqueous Volume: 115 microliter, Exterior: 12.5L x 12.5W x 45mmH, Window: 2W x 5mmH, Z-Dimension: 15mm