Learn More
Starna Cells Inc SUB MICRO CELL 100UL
SDP
Catalog No. NC9982880
Encompass_Preferred
Sub-Micro Fluorometer Cells, Type: 16F, Path Length: 10mm, Exterior: 12.5L x 12.5H x 45mm W, Window: 2W x 5mm H, Z Dimensions: 20mm, Geometric Volume: 100microliter, Aqueous Volume: 115microliter.
Catalog No. NC9982880 Supplier Starna Cells Inc Supplier No. 16.100Q10/Z20
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Sub-Micro Fluorometer Cells, Type: 16F, Path Length: 10mm, Exterior: 12.5L x 12.5H x 45mm W, Window: 2W x 5mm H, Z Dimensions: 20mm, Geometric Volume: 100microliter, Aqueous Volume: 115microliter.