Learn More
Starna Cells Inc SUB MICRO CELL QUARTZ 100UL
SDP
Catalog No. NC9970140
Encompass_Preferred
Sub-Micro Cell, Square top, two translucent walls, Spectrosil quartz, 2W x 5H, 12.5L x 12.5W x 45H, 0.100mL, Open top, supplied with non-sealing PTFE cover as well as a vaned lid to provide a liquid-tight seal
Catalog No. NC9970140 Supplier Starna Cells Inc Supplier No. 16.100Q10/Z8.5
Sub-Micro Cell, Square top, two translucent walls, Spectrosil quartz, 2W x 5H, 12.5L x 12.5W x 45H, 0.100mL, Open top, supplied with non-sealing PTFE cover as well as a vaned lid to provide a liquid-tight seal